Would you like to log in again?
Mobile Wire Security Units, Metro™
Supplier: Metro International
SEC53EC SEC63S-SD SEC56ECQ SEC53ECQ SEC66EC SEC53ECQ SEC55DCQ SEC53DCQ SEC56S-HD SEC56EC SEC53S-SD SEC53DK3 SEC55EK3 SEC35S-SD SEC63EC SEC56DCQ SEC33EC SEC56DK3 SEC65S-SD SEC53DK3 SEC35EK3 SEC33S-SD SEC55S-SD SEC35EK3 SEC55DCQ SEC55S-HD SEC53S-HD SEC33S-SD SEC56EC SEC56EK3 SEC66EC SEC55ECQ SEC63S-SD SEC56EK3 SEC65S-SD SEC55S-HD SEC55DK3 SEC53DCQ SEC56ECQ SEC56S-SD SEC65EC SEC56DK3 SEC66S-SD SEC53EK3 SEC35S-SD SEC55EC SEC63EC SEC35ECQ SEC55S-SD SEC53S-HD SEC33ECQ SEC66S-SD SEC65EC SEC56DCQ SEC53S-SD SEC56S-HD SEC35ECQ SEC53EK3 SEC33EK3 SEC53EC SEC55DK3 SEC55EK3 SEC35EC SEC33ECQ SEC33EK3 SEC56S-SD SEC35EC SEC55ECQ SEC33EC SEC55EC
82023-466EA 5192.18 USD InStock
CA22233-696 CA22233-698 CA82023-430 CA82023-474 82023-466 82023-422 CA82023-476 CA82023-432 82023-460 CA82023-472 CA82023-426 CA82023-428 CA82023-506 CA82023-422 CA82023-466 82023-458 CA82023-424 CA82023-502 CA82023-440 82023-476 82023-432 CA82023-486 CA82023-442 82023-434 82023-472 CA82023-482 82023-474 82023-430 82023-428 CA82023-438 82023-506 82023-424 CA82023-434 82023-502 82023-426 CA82023-436 CA82023-490 22233-700 82023-486 82023-442 82023-444 CA82023-498 82023-482 82023-440 CA82023-494 CA82023-450 CA82023-448 97005-776 97005-778 CA82023-444 82023-436 CA82023-446 82023-438 22233-698 82023-490 22233-696 CA22233-700 82023-498 82023-456 82023-450 82023-494 CA82023-460 97005-774 CA97005-774 CA97005-776 82023-446 CA97005-778 CA82023-456 82023-448 CA82023-458
Mobile Wire Security Units, Metro™
Carts
Security units are designed to protect materials and sensitive items from loss or pilferage
Note: Given height is for unit without a Metro dolly or casters.
Units move easily to allow thorough cleaning or inspection, and they provide the advantage of quick, protective transportation. Heavy-gauge, open wire construction allows contents to be visible at all times, making inspection and retrieval quick and simple. An ergonomic, 1/4-turn door handle simplifies opening and closing. Each unit is equipped with a set of double doors that open 270° and can be secured along the sides of the unit. Super Erecta* and qwikSLOT* shelf designs are easily adjustable and allow for flexibility to meet changing needs. Super Erecta shelves attach to posts via tapered sleeves; qwikSLOT shelves rest securely on support clips that can be quickly and easily repostitioned. Shelves can be positioned in 2.5cm (1") increments along the entire height of the posts. Security units are available in chrome, stainless steel, or Metroseal 3, which provides Microban* antimicrobial product protection. All units are shipped knocked-down and can be quickly and easily assembled right from packaging.
Ordering information: Chrome and Metroseal 3 units are supplied with 12.7cm (5") standard-duty polyurethane-tread stem casters. Stainless steel units do not include casters; see 19766-236 series to purchase them. Contact your Avantor representative for the proper caster for your application. For all units, interior shelves must be purchased separately. For Super Erecta shelves, see 22233-716 series; for qwikSLOT shelves, contact your Avantor representative. For stationary wire security units, see 82023-452 series. Heavy-duty stainless steel models include staked posts for use with standard Metro dollies. Dolly bases and plate casters must be ordered separately; contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty