Would you like to log in again?
SmartWall Plus™ Grids and Shelving Accessories, Metro™
Supplier: Metro International
SmartWall Plus™
WG1836K3 FGS1224K3 WG1860K3 PGHK6K3 GS1848K3 WG1848K3 WG2436K3 GS1848K3 GS1436K3 WG3036K3 BH6K3 WG2448K3 WG1848K3 GS1436K3 WG2448K3 GS1830K3 WG2436K3 BH6K3 GS1836K3 GS1448K3 WG3036K3 FGS1224K3 WG1836K3 PGHK6K3 WG3048K3 WG1860K3 GS1448K3 WG3048K3 GS1836K3 GS1830K3
83009-610EA 115.68 USD InStock
83009-610 83009-612 83009-614 82023-620 CA82023-630 CA82023-624 82023-616 CA82023-626 82023-618 CA82023-620 82023-634 CA82023-622 82023-614 82023-636 CA82023-628 CA82023-608 82023-630 82023-632 82023-626 CA82023-614 CA82023-636 82023-628 CA82023-616 82023-622 CA82023-632 CA82023-610 82023-624 CA82023-634 CA82023-612 CA82023-618
SmartWall Plus™ Grids and Shelving Accessories, Metro™
Graticules, Reticles and Grids
Grids and shelving accessories allow users to organize SmartWall Plus storage systems to precise specifications.
  • Grids maintain a total unit weight capacity of 114 kg (250lbs.)
Storage options can be easily repositioned along the track before securing them to the wall to maximize storage space and improve organization. Metroseal three finishes provide Microban® built-in product protection.
Ordering information: Accessory bracket kits for wire grids (82023-344) include two top brackets that attach to track and four bottom brackets that attach to wall surface. One kit is required per grid. Brackets are constructed of Type 304 stainless steel. Bracket kits for Starsys® wall cabinets (82023-334) include two top brackets and one bottom bracket. Top brackets are Type 304 stainless steel and bottom brackets are steel coated with corrosion-resistant taupe epoxy.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty