Would you like to log in again?
VWR® Gas Cylinder Wall Mounts and Supports
Supplier: VWR
VWR®
4302808 4302809 4302807 4302806 4302811 4302810
82023-836EA 962.52 USD InStock
82023-836 82023-826 82023-832 82023-834 82023-830 82023-828
VWR® Gas Cylinder Wall Mounts and Supports
Wall Hooks
Cylinder wall brackets secure 10.2 – 30.5 cm (4 – 12") diameter cylinders using 137.2L x 3.8Wcm (54 x 11/2") polypropylene straps that can withstand up to 1200psi.
  • Polypropylene strap
  • Durable brackets and mounts
The single cylinder adjustable bracket permanently mounts to wall to secure one 10.2 – 35.6 cm (4 – 14") diameter cylinder using a polypropylene strap and security chain. The bench mount bracket holds one 10.2 – 35.6 cm (4 – 14") diameter cylinder using dual mounting posts with swivel feet and nonslip pads to clamp to benches or tabletops from 1 – 4.4 cm (3/8 – 13/4") thick. The cylinder floor stand safely supports one 10.2 – 25.4 cm (4 – 10") diameter cylinder.
Ordering information: Fasteners are not included; contact your VWR® sales representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty