Would you like to log in again?
VWR® Digital Stopwatch
Supplier: VWR
61700-5311
82023-864CS 1116.31 USD InStock
82023-864
VWR® Digital Stopwatch
Timers Digital Stopwatches
Supplied with multi-language (English, French, German, Italian, Portuguese, and Spanish) instructions, this electronic stopwatch includes a calendar and a clock.
  • Features Certificate of Calibration and individual serial number
  • Will store 1 lap time and includes an alarm
  • Requires one CR2032 battery (included)
Certifications: Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty