Would you like to log in again?
Test Tubes without Rim
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TT9820-H TT9820-E TT9820-D TT9820-C TT9820-B TT9820-I TT9820-K TT9820-L TT9820-A TT9820-F TT9820-J
82024-106CS 695.9 USD InStock
82024-106 76532-222 76532-234 82024-104 76532-220 76532-232 76532-226 76532-224 76532-236 76532-228 76532-230
Test Tubes without Rim
Tubes Test Tubes
Pre-cleaned borosilicate glass test tubes without rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes do not have rims
  • Autoclavable
Rimless borosilicate glass test tubes are ideal for a variety of laboratory applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty