Would you like to log in again?
Dropper® Urine Chemistry Controls, Quantimetrix
Supplier: Quantimetrix
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Dropper®
1432-31 1431-31
82024-154BX 144.25 USD InStock
82024-154 82024-156
Dropper® Urine Chemistry Controls, Quantimetrix
Clinical Diagnostic Supplies Clinical Reagents and Kits
Use Dropper Urine Chemistry Control to monitor the performance of routine and stat urine chemistry assay methods. Drop the control directly into the instrument sample cup. The easy-to-use dropper bottle provides simple dispensing and maximum use.
  • Easy-to-use dropper bottles
  • Liquid, made with human urine
  • 2-year shelf life and open vial stability from date of manufacture when stored at 2 to 8°C
Dropper Urine Chemistry Control is designed for use with most major chemistry analyzers including Siemens Advia®, Beckman Coulter AU® Series, Siemens Dimension®, Beckman Coulter Synchron®, Ortho Vitros®, Abbott Architect and Roche Modular, as well as for the Pyrogallol Red microprotein method and osmolality (freezing point method).
Ordering information: Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty