Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trypan blue 0.4% in PBS

Supplier: Cytiva
Synonyms: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium saltCI 23850Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulfonate)Direct Blue 14AzidinBlueSodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulfonic acid tetrasodium saltBenzamine blueTrypan blue sodium salt3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulfonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt

SV30084.01 SV30084.01
82024-256EA 32.3 USD InStock
CA82024-256 82024-256
Trypan blue 0.4% in PBS
Trypan Blue
Trypan Blue solution, 0,4%, is routinely used as a cell stain to assess cell viability using the dye exclusion test.


Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
MDL Number: MFCD00003969
CAS Number: 72-57-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty