Would you like to log in again?
HyClone™ SG-200 Solution, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30590.01 SH30590.01
82024-258EA 37 USD InStock
CA82024-258 82024-258
HyClone™ SG-200 Solution, HyClone products (Cytiva)
Cell Culture Supplements and Additives
HyClone SG-200 Solution is a stable dipeptide of L-Alanyl L-Glutamine
Storage conditions: Store at 2° to 8°C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty