Would you like to log in again?
SMS Hoodless Coverall, International ENVIROGUARD™
Supplier: International Enviroguard
2223-5XL
82024-312CS 185.74 USD InStock
82024-312
SMS Hoodless Coverall, International ENVIROGUARD™
Disposable Garments
Coverall is designed for applications in industrial facilities, laboratories, research and clinical facilities, pharmaceutical facilities, food processing facilities, and construction and abatement sites
Note: It is the responsibility of the user to select garments which are appropriate for intended use and which meet all health, safety, and regulatory requirements.
Coverall is constructed of a three-layer, spunbond/meltblown/spunbond material. It provides some splash resistance, strength, and mobility in a lightweight fabric while maintaining comfort, breathability, and softness. Coverall features elastic wrists, ankles, and back, as well as a zipper cover. Weight: 60g (23.5oz.).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty