Would you like to log in again?
VWR® Slide Mailers for 5 Slides
Supplier: VWR
VWR®
HS15983GV HS15982V
82003-431CS 713.99 USD InStock
82003-431 82024-598
VWR® Slide Mailers for 5 Slides
Slide Mailers
Ideal for shipping, storing, or staining slides, these polypropylene containers hold five standard 25x75 mm (1x3") slides.
Inside wall features slotted channels to keep slides separated and make removal easier. Closes securely to keep slides safe.
Ordering information: Assorted colors include red, yellow, blue, purple, and orange.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty