Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Penicillin/Streptomycin/Fungizone solution 100X, HyClone™

Supplier: Cytiva

SV30079.01 SV30079.01
82026-730EA 49.85 USD InStock
CA82026-730 82026-730
Penicillin/Streptomycin/Fungizone solution 100X, HyClone™
Penicillin/Streptomycin/Fungizone
Antibiotic-Antimycotic (Pen/Strep/Fungiezone) Solution is used to control or destroy destructive microorganisms, such as gram negative, gram positive bacteria, fungi and yeast.

  • Contains 10 000 units of penicillin, 10 000μg of streptomycin, and 25 μg of Amphotericin B per ml
  • Wide spectrum of activity against gram negative and gram positive bacteria, as well as activity against fungi and yeast
  • Commonly used as an anti-fungal, anti-yeast, anti-mould or anti-bacterial
  • Common working dose is 10 ml/l of 100X solution


Order Now
Specification Test Results
Format Liquid
Storage Store frozen
Antibiotic Spectrum Broad spectrum antibacterial and antifungal
Concentration 100X
Shelf life 18 months
Sterilization 0.2 µm sterile filtered

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty