Would you like to log in again?
Upchurch Scientific® Manifolds, Multi-Port Connectors, IDEX Health & Science
Supplier: IDEX Health & Science
Upchurch Scientific®
P-155 P-150
82026-908EA 215.39 USD InStock
82026-908 82026-910
Upchurch Scientific® Manifolds, Multi-Port Connectors, IDEX Health & Science
Fittings and Connectors
Upchurch Scientific® Standard Manifolds from IDEX Health & Science are available in 5, 6, 7 or 9 Port configurations to combine several streams into one, or split one fluid stream into several
Each PEEK™ manifold comes complete with 1/4-28 Super Flangeless™ Fittings for either 1/16 in or 1/8 in OD tubing, with pressure ratings of 2,000 psi (138 bar) and 500 psi (34 bar), respectively. Ideal manifold applications include: Multiport mixing chamber, gas sparging splitting union, and sample injection onto multi-well plates or multiple direction flow path unions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty