Would you like to log in again?
VWR® Crucible Tongs
Supplier: VWR
VWR®
11648-104 11648-101
82027-362CS 925.11 USD InStock
82027-362 82027-368
VWR® Crucible Tongs
Tongs
Crucible tongs are designed for use in handling a wide variety of laboratory apparatus.
  • Nickel-plated stainless steel fabrication
The 24.1cm model is a general-purpose tong with serrated tips. The 45.7cm model features nickel-plated steel construction and an extra-long design.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty