Would you like to log in again?
VWR® Beaker Tongs, Fiberglass Coating
Supplier: VWR
VWR®
11648-107
82027-374CS 2484.79 USD InStock
82027-374
VWR® Beaker Tongs, Fiberglass Coating
Tongs
Tongs feature a stainless steel construction, a riveted joint, and fiberglass coating for use in high-temperature applications.
  • Stainless steel construction
  • For use in high-temperature applications
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 mL capacities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty