Would you like to log in again?
VWR® Hemostatic Forceps
Supplier: VWR
VWR®
11648-127 11648-126
82027-414CS 1201.42 USD InStock
82027-414 82027-412
VWR® Hemostatic Forceps
Forceps
Hemostatic forceps are used in many surgical procedures, usually to control bleeding or the flow of liquids in tubing.
  • Stainless steel fabrication
These forceps have ring handles just like scissors, and ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty