Would you like to log in again?
VWR® Rochester-Pean Hemostatic Forceps
Supplier: VWR
VWR®
11648-130 11648-132 11648-129 11648-131
82027-418CS 2183.71 USD InStock
82027-418 82027-424 82027-420 82027-422
VWR® Rochester-Pean Hemostatic Forceps
Forceps
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
  • Stainless steel construction
  • Reusable
  • Non-sterile
  • Premium grade
Horizontal serrations help ensure forceps remain clamped tight.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty