Would you like to log in again?
VWR® Gooseneck Forceps
Supplier: Avantor
VWR®
11648-142
82027-444CS 1064.65 USD InStock
82027-444
VWR® Gooseneck Forceps
Forceps
These stainless steel forceps are used for dissecting, handling insects, or positioning specimens for pinning.
  • Stainless steel construction
  • Serrated tips
The forceps have serrated tips and a handle with a gooseneck shank.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty