Would you like to log in again?
VWR® Smooth Jaw Forceps
Supplier: Avantor
VWR®
11648-144
82027-448CS 1621.99 USD InStock
82027-448
VWR® Smooth Jaw Forceps
Forceps
Stainless steel with straight, smooth jaws are ideal for use when you need an extra pair of hands on any number of craft applications.
  • Stainless steel construction
  • Straight, smooth jaws
  • Adjustable tension
  • Quick release
Smooth jaws will not make any mark on fabric.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty