Would you like to log in again?
VWR® Self-Closing Forceps
Supplier: Avantor
VWR®
11648-146
82027-452CS 320.56 USD InStock
82027-452
VWR® Self-Closing Forceps
Forceps
Stainless steel self-closing forceps feature straight, fine tips and a smooth grip.
  • Stainless steel construction
  • Fine tips
  • Self-closing design
Forceps are passivated and autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty