Would you like to log in again?
VWR® PTFE-Coated Forceps
Supplier: VWR
VWR®
11648-147
82027-454CS 2261.22 USD InStock
82027-454
VWR® PTFE-Coated Forceps
Forceps
Stainless steel forceps are PTFE-coated.
  • Stainless steel construction
  • Chemically resistant
  • Nonsticking and non-corroding
Forceps are coated with PTFE to imbue forceps with heat and chemical resistance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty