Would you like to log in again?
VWR® Spatula, PTFE-Coated
Supplier: Avantor
VWR®
11648-187
82027-534CS 1216.11 USD InStock
82027-534
VWR® Spatula, PTFE-Coated
Spatulas
Stainless steel spatula features one rounded end and one flat end, with PTFE coating throughout.
  • Stainless steel construction
  • PTFE coating
  • Nonsticking and non-corrosive
It can be used to transfer corrosive or sticky materials without scratching or damaging container surfaces.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty