Would you like to log in again?
ColorZip™ Reclosable Storage Bags
Supplier: Minigrip
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ColorZip™
MGDZW270Q MGDZW175G MGDZW270G MGDZW175J MGDZW175Q MGDZW270J
82027-626CS 63.56 USD InStock
82027-626 82027-628 82027-634 82027-636 82027-630 82027-632
ColorZip™ Reclosable Storage Bags
Bags Storage Bags
Reclosable polyethylene bags feature easy open/close zipper and external grip strips for simplified handling.
  • Single zipper and white write-on block standard for sandwich size
  • Double zipper and white write-on block is standard for quart, gallon and jumbo storage and freezer bags
  • Double zipper feature provides greater leak-resistance
  • Freezer grade is thick and heavy duty
  • All bags feature a white write-on block
Large offset lip with external grip ridges for easy handling, even with gloves. Storage bags have improved cold-fracture resistance for short freezing (1 to 2 weeks).
Certifications: FDA approved for contact with food.
Packaging: Crush-resistant dispenser cartons offer protection in the toughest environments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty