Would you like to log in again?
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR
VWR®
89523-484A 82028-428A 82028-426A 82028-432A 89523-482A
89523-484ACS 808.18 USD InStock
89523-484A 89523-482A 82028-428A 82028-426A 82028-432A
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Vials
10-425 autosampler screw thread vials feature a wide opening compatible with 6 mm inserts.
  • Choose from all-steel or improved polymeric-metal version
  • Both compatible with all standard or magnetic Headspace autosamplers
  • Excellent sealing characteristics that withstand high pressures of headspace applications
Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty