Would you like to log in again?
Nalgene® Biotainer® Polycarbonate Carboys, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® Biotainer®
3405-42 3423-42 3410-42 3233-42 3410-08 3405-16
82028-472CS 222.74 USD InStock
82028-472 82028-462 82028-476 82028-466 82028-474 82028-464
Nalgene® Biotainer® Polycarbonate Carboys, Thermo Scientific
Bottles Carboys
Sterile carboys are molded from blue-tinted polycarbonate and provide safe storage for contents at temperatures from −85 °C (−121 °F) to near boiling
  • Space-saving square shape
Carboys meet USP-VI and are noncytotoxic and nonpyrogenic. Gamma irradiated to a sterility assurance level of 10⁻⁶.
Ideal for storage and transport of media, sera, antigens, and other biological reagents. Carboys feature molded graduations in liters and gallons and indented, ribbed handgrips.
Ordering information: Each carboy is supplied with a 48mm screw cap with a silicone liner. Individually bagged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty