Would you like to log in again?
Nalgene® InVitro™ Biotainer Bottles, Sterile, PETG, Thermo Scientific
Supplier: Thermo Scientific
3110-42 3005-70 3005-42 3110-35 3025-42 3230-42 3230-20
82028-490CS 560.64 USD InStock
82028-490 82028-480 82028-484 82028-482 82028-488 82028-486 82028-478
Nalgene® InVitro™ Biotainer Bottles, Sterile, PETG, Thermo Scientific
Bottles Plastic Bottles
Nalgene® InVitro™ Biotainer Bottles are designed specifically for bio-pharmaceutical manufacturers.
The bottles are made from crystal clear PETG, feature a space-saving square shape with ribbed hand grips (except 125 ml), and the closures feature a polyethylene foam core liner.
They reduce the risk of carry-over contamination by eliminating the cost of cleaning, sterilizing and associated validation.
Certifications: Meet USP 87, 88. Lot tested for pyrogenicity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty