Would you like to log in again?
Nalgene® Invitro™ Biotainer Bottles, Sterile, HDPE, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
3751-42 3750-24 3751-24
82028-494CS 743.04 USD InStock
82028-494 82028-492 82028-496
Nalgene® Invitro™ Biotainer Bottles, Sterile, HDPE, Thermo Scientific
Bottles Plastic Bottles
Nalgene® InVitro Biotainer bottles reduce the risk of carryover contamination and eliminate the cost of cleaning, sterilizing and its associated validations.
  • The bottles are supplied sterile and ready to use, and offer excellent chemical resistance
They are made from HDPE with a silicone-lined polypropylene 38 mm closure. They feature printed graduations to 3000 ml, with a 3900 ml brim capacity and a space-saving square shape with a convenient handle.
Certifications: Meet USP 87, 88. Lot tested for pyrogenicity.
Ordering information: 3750-24 bottles feature tamper-evident shrink bands around each neck and closure.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty