Would you like to log in again?
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport
SLIMSETTE™
M509-12 M509-16 M509-16 M509-3 M509-7 M509-10 M509-6 M509-2 M509-2 M509-10 M509-5 M509-9 M509-17 M509-8 M509-18 M509-18 M509-5 M509-4 M509-11 M509-12 M509-17 M509-6 M509-4 M509-11 M509-9 M509-3
82028-758CS 465.13 USD InStock
CA82028-768 CA82028-746 CA82028-766 CA82028-744 82028-758 CA82028-748 CA82028-760 82028-752 82028-750 CA82028-764 CA82028-762 82028-770 CA82028-758 82028-748 82028-746 82028-768 CA82028-756 CA82028-750 82028-762 82028-760 CA82028-770 82028-766 CA82028-754 82028-744 82028-764 CA82028-752
Slimsette™ Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
These compact, easy-to-use cassettes incorporate a unique recessed cover which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1x 5mm slotted openings ensure efficient fluid exchange and drainage, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle
Certifications: CE marked.
Ordering information: Packaged in three dispenser boxes of 500 cassettes with covers assembled.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty