Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Disposable Carbon Steel Surgical Blades, OR Grade, Sklar®
Supplier: Sklar
06-3012 06-3015 06-3020 06-3021 06-3011 06-3023 06-3022 06-3010
82029-822PK 115.38 USD InStock
82029-822 82029-824 82029-826 82029-828 82029-818 82029-830 82029-820 82029-832
Disposable Carbon Steel Surgical Blades, OR Grade, Sklar®
Scalpels and Scalpel Assemblies Scalpel Blades
Disposable Surgical Blades are carbon steel, industry standard
Commonly used for large, straight incisions. Blades feature a curved cutting edge. Supplied sterile in a box of 100.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty