Would you like to log in again?
Lister Bandage Scissors, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11-1035C 11-1045C 11-1045 11-1080C 11-1055C 11-1072 11-1055 11-1072C 11-1060 11-1080 11-1060C 11-1035
82029-878EA 170.87 USD InStock
82029-878 75847-018 82029-866 75847-016 82029-868 75847-014 75847-012 75847-022 82029-870 75847-020 82029-872 82029-874
Lister Bandage Scissors, Sklar
Scissors
Lister Bandage Scissors are a versatile instrument with multiple uses. They are commonly used for sizing dressings and removing circumferential bandages, but may also be used to cut tougher material such as plaster, fabric, andoccasionally umbilical cord.
  • Scissors are autoclavable
  • Product is constructed with premium grade stainless steel
  • 82029-872 feature a large ring
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer.
The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without cutting the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Ordering information: The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty