Would you like to log in again?
Littauer Stitch Scissors, Sklar
Supplier: Sklar
22-1354 15-1355
89025-248EA 198.73 USD InStock
89025-248 82029-942
Littauer Stitch Scissors, Sklar
Scissors
Sklar's® Littauer Stitch Scissors are primarily used for suture removal.
  • Small hook-shaped tip on one blade
  • Larger, less delicate tip for removing larger sutures
These scissors have a small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer Stitch Scissors have larger, less delicate tips than Spencer Stitch Scissors, and are used for removing larger sutures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty