Would you like to log in again?
Mayo Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
15-2555 15-1555 90-1590 15-1567 90-1690 15-2567
82029-944EA 103.92 USD InStock
82029-944 10192-384 82029-946 82029-958 10192-314 82029-960
Mayo Dissecting Scissors, OR Grade, Sklar
Scissors
Mayo Dissecting Scissors are a versatile instrument, that can be used in multiple procedures and settings.
They are available straight or curved, in a variety of lengths, and have a standard beveled blade
Censitrac Ready products are industry standard and constructed of stainless steel.
While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty