Would you like to log in again?
Halsted Mosquito Hemostatic Forceps, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
17-1220 17-1225 17-1550 17-1450
82029-970EA 82.99 USD InStock
82029-970 82029-962 82029-972 82029-964
Halsted Mosquito Hemostatic Forceps, Sklar
Forceps
Halsted Mosquito Forceps are made of high-quality stainless steel and these forceps are used in multiple procedures as a hemostat for small blood vessels. Mosquito forceps are known for their fine tips, and short, fully serrated jaws.
  • Autoclavable and reusable
  • Non-sterile and latex-free
  • Available curved or straight
  • Ratcheted finger ring handle provides a secure, locking grip to halt blood flow
  • Not appropriate for use with larger vessels or arteries
A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty