Would you like to log in again?
Rochester-Pean Hemostatic Forceps, Sklar
Supplier: Sklar
17-2462 17-2455 17-2262 17-2472C 17-2255 17-2262C 17-2472 17-2455C 17-2462C 17-2480C 17-2272C 17-2255C 17-2280C 17-2272 17-2480 17-2280
82029-988EA 108.72 USD InStock
82029-988 82029-978 75847-168 75847-178 75847-166 75847-176 82029-980 82029-990 82029-982 82029-992 82029-984 82029-986 75847-174 75847-172 75847-170 75847-180
Rochester-Pean Hemostatic Forceps, Sklar
Forceps
Sklar® Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
  • Available curved or straight
  • Latex-free
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures
Censitrac Ready OR Grade Rochester-Pean Forceps are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty