Would you like to log in again?
St. Vincent Tubing Forceps, OR Grade, Sklar
Supplier: Sklar
17-3962
82030-020EA 157.61 USD InStock
82030-020
St. Vincent Tubing Forceps, OR Grade, Sklar
Forceps
Sklar's® St
Censitrac Ready products are industry standard and constructed of stainless steel.
Vincent tubing forceps is a ratcheted, finger ring instrument used to block the flow of fluids from large chest tubes or tubing from bypass machines.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty