Would you like to log in again?
Standard Fine Splinter Forceps, OR Grade, Sklar
Supplier: Sklar
19-3045 19-3040
82030-086EA 40.57 USD InStock
82030-086 82030-084
Standard Fine Splinter Forceps, OR Grade, Sklar
Forceps
Sklar® Plain Splinter Forceps are multi-purpose thumb forceps typically used for grasping and removing small foreign objects with their fine and pointed tips
It is also used for holding bandages, sutures, or other items.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty