Would you like to log in again?
Halsey Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-1751
82030-110EA 183.39 USD InStock
82030-110
Halsey Needle Holder, OR Grade, Sklar®
Forceps
Sklar® Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology.
They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty