Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Mayo-Hegar Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2052 20-2911 20-2070 20-2490 20-2044 20-2048 20-2046 20-2270 20-2060 21-2570 21-2480 20-2910 20-2480
82030-118EA 156.07 USD InStock
82030-118 82030-116 82030-114 82030-112 82030-122 76192-504 10195-050 76192-502 82030-120 89023-096 76192-500 89023-094 10195-048
Mayo-Hegar Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument for multiple procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty