Would you like to log in again?
Gelpi Retractor, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22-8855 22-8853 60-6570 22-8851 22-8850 60-6580
10197-828EA 559.33 USD InStock
10197-828 82030-158 82030-156 76192-932 82030-154 10197-826
Gelpi Retractor, OR Grade, Sklar®
Retractors
The Sklar® Gelpi Retractor is a ratcheted, finger ring retractor often used in smaller surgical sites.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
A single sharp prong on each tip is turned outward, sideways, at a 90° angle. Once the targeted tissue has been held back, the locking mechanism allows the retractor to remain in place, and does not require someone to hold it during the procedure.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty