Would you like to log in again?
Iris Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
47-1244 64-2042 47-1135 22-1235 47-1243 47-1240 47-1248 22-1135 47-1440 47-1145 64-2142 47-1246 47-1140 47-1144 47-1441 47-1143 47-1442 47-1235 47-1245 64-2143
89025-212EA 154.77 USD InStock
89025-212 82030-238 82030-246 89025-228 82030-236 82030-244 82030-234 82030-242 10194-364 82030-232 82030-240 82030-230 10194-356 10194-358 89024-952 89024-964 89024-962 10194-360 10197-872 10194-362
Iris Scissors, OR Grade, Sklar
Scissors
Iris Scissors are designed for ophthalmic procedures, these fine and sharp scissors are suitable for detailed dissection of fine tissue and fine suture removal.
  • Multipurpose instrument used in various surgical environments
In addition to ophthalmic procedures, Iris scissors are used in emergency rooms and for obstetrics and gynecology as well as dermatologic procedures.
The Ready OR Grade Iris Scissors are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty