Would you like to log in again?
McPherson Vannas Scissors, Sklar
Supplier: Sklar
64-1961 64-1960
82030-296EA 718.26 USD InStock
82030-296 82030-294
McPherson Vannas Scissors, Sklar
Scissors
McPherson Vannas Scissors are double spring thumb scissors commonly used in ophthalmic procedures such as iridotomy.
These scissors feature 10 mm, sharp, delicate blades ideal for use on fine tissues of the eye
Censitrac Ready products are industry standard and constructed of stainless steel.
The handle has a horizontally serrated grip that provides greater control of the instrument.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty