Would you like to log in again?
Jewelers Style Forceps, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
66-7542 66-7642 66-7442 66-7645 66-7447 66-7644 66-7436 66-7438 66-7440
82030-316EA 59.75 USD InStock
82030-316 82030-328 82030-314 82030-312 82030-324 82030-322 82030-330 82030-320 82030-318
Jewelers Style Forceps, Sklar
Forceps
Jeweler Style Forceps are grasping forceps used in microsurgical procedures
  • Non-Sterile
  • Latex-Free
  • Autoclavable and reusable
  • Solid knurled handle
They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. These multi-purpose thumb forceps with flat handles are available with various tip styles.
Swiss-style nonmagnetic forceps are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty