Would you like to log in again?
Puritan® Large Flat Paddle Shaped Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
3600 3600
82030-704CS 7303.84 USD InStock
CA82030-704 82030-704
Puritan® Large Flat Paddle Shaped Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
For use in critical environment cleaning
Chemical-resistant, nonabrasive, symmetrical tip is free from volatile residues, particles, contaminating adhesives, and lint. Polypropylene shaft. Made in the USA.
Certifications: CE marked.
Ordering information: Swabs are packaged 50 per pack, 20 packs per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty