Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rodent RQ 22-9 Diet, Zeigler™

Supplier: Zeigler
413000-75-53
82031-012EA 68.21 USD InStock
82031-012
Rodent RQ 22-9 Diet, Zeigler™
Animal Supplies Animal Feed
Formulated for reproduction and lactation of both rats and mice, and strains of rodents that have higher energy requirements

  • Recommended for use in production and research rodent colonies
  • Contains 22% protein, 9% fat
  • Wafer pellet size
  • Diet supplied in 22.7 kg (50 lbs.) bag

Ingredients include: wheat, corn, soybean meal, wheat middlings, wheat flour, soy oil, fish meal, corn gluten meal, alfalfa meal, brewer’s dried yeast, dicalcium phosphate, ameribond, salt, limestone, choline chloride, d-activated animal sterol (source of vitamin D3), dl-alpha tocopheryl acetate (vitamin E), folic acid, niacin, riboflavin, thiamine mononitrate, vitamin B12 supplement, pyridoxine hydrochloride, biotin, vitamin A acetate, menadione sodium bisulfite complex (vitamin K), calcium pantothenate, cobalt carbonate, copper sulfate, ferrous sulfate, and manganese oxide.

Max. ash: 7%
Max. crude fibre: 4%

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty