Would you like to log in again?
RABBIT NIH 09 165 24 30LB
Supplier: ZEIGLER BROS. INC.
Rabbit NIH-09 16.5-2.4 Fixed Formula Lab Diet^ Designed for growth, maintenance, and reproduction of rabbits; 16.5% Protein, 2.4% Fat; 16.8% Fiber; 5/32" pellet size, 30lb bag.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RABBIT NIH 09 165 24 30LB 82031-042 410900-53-30 Each (30lb) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty