Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit NIH-09 Open Formula, Zeigler™

Supplier: Zeigler
410900-53-53
82031-044EA 37.11 USD InStock
82031-044
Rabbit NIH-09 Open Formula, Zeigler™
Animal Supplies Animal Feed
Formulated for maintenance, growth, and reproduction of conventional rabbits

  • Contains 16.5% protein, 2.4% fat
  • Pellet size: 4mm (5/32")
  • 22.7 kg (50 lbs.)

Ingredients include: ground barley, soybean meal, oat hulls, wheat middlings, wheat bran, dired whey, alfalfa meal, brewer’s dried yeast, soy oil, dried molasses, dicalcium phosphate, limestone, salt, and premixes.

Max. ash: 10%
Max. crude fibre: 16.8%

Certifications: Used at the National Institutes of Health (NIH) as the standard reference diet for rabbit production and in rabbit colonies involved in biological and biomedical research.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty