Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

11 mm Standard Opening Crimp-Top Vials, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CERT4011-89W C4012-2 C4012-2W C4012-88A C4012-1 C4012-15 C4012-1W
82031-698PK 51.51 USD InStock
82031-698 66065-128 66030-244 66030-242 66030-248 89026-588 66011-240
11 mm Standard Opening Crimp-Top Vials, Thermo Scientific
Vials
Crimp-top vials have a narrow neck compatible with 11 mm aluminum seals and 5 mm inserts.

Inserts are made of clear, borosilicate glass. These inserts are custom fitted for use with standard opening, 2 ml, 12×32 mm vials. Flat-bottom construction optimizes the depth of the vial.

The conical cavity allows maximum retrieval of contents with a syringe. All vials are manufactured from borosilicate glass, except for Glastic® plastic vials, which have a fixed glass 150 µl insert. Glass MicroVial™ vials are for limited volume applications.

Certifications: Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty