Would you like to log in again?
ThinCert™ Tissue Culture Inserts, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ThinCert™
657631 662641 657630 662631 657610 662610 665610 657638 665640 662638 665630 662630 665641 657641 662640 657640 665631 665638
82051-572CS 194.86 USD InStock
82051-572 82051-570 82051-568 82050-026 82050-016 82050-038 82050-028 82050-018 82050-030 82050-020 82050-032 82050-010 82050-022 82050-034 82050-012 82050-024 82050-014 82050-036
ThinCert™ Tissue Culture Inserts, Sterile, Greiner Bio-One
Microplates Insert Plates and Inserts
Tissue culture inserts provide optimal support for co-cultivation, cell migration, and transport process applications.
  • Sterile
  • Spacers eliminate capillary effect
  • Individually wrapped inserts
  • Low protein binding capacity
  • Improved cell adhesion
Advanced design permeable inserts feature transparent or translucent polyethylene terephthalate (PET) membranes that offer low protein binding capacity, improved cell adhesion, and defined pore size to ensure complete exchange of compounds and nutrients
ThinCert™ membranes are sealed to a robust, transparent polystyrene housing. Housing features self-positioning hanging geometry with eccentric in-well positioning, improving both the pipetting angle and window. Spacers maintain distance to eliminate capillary effect. Inserts are individually wrapped.
Membrane will not curl after detachment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty