Would you like to log in again?
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cryo.s™
127279 126279 127278 122277 122278 126277 124274 121279 127261 124275 126278 124273 126280 123280 121278 126261 121280 123263 121263 127280 122261 127277 121261 123278 124276 122280 124261 127263 122263 122279 123261 121277 123279 126263 124263 122272 123277
82050-190CS 181.44 USD InStock
82050-190 10799-204 82050-170 82050-192 82050-158 82050-214 82050-216 82050-218 82050-172 82050-194 82050-196 82050-174 82050-198 82050-154 82050-176 82050-210 82050-156 82050-178 82050-212 82050-180 82050-160 82050-182 82050-224 82050-204 82050-226 82050-206 82050-208 82050-162 82050-184 82050-164 82050-186 82050-220 82050-188 82050-166 82050-222 82050-200 82050-168
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Vials
Designed for short- and long-term storage of samples and cells, these cryogenic storage vials are made from virgin polypropylene resin for purity, clarity, and consistency.
  • Nonpyrogenic and RNase- and DNase-free to <0.06 EU/ml
Cryo.s™ vials are available with internal or external thread. The internal thread format features a silicone gasket and is ideally suited for storage in the vapor phase of liquid nitrogen, at temperatures from –196 to 121 °C.
Delivery information: All vials are supplied with caps. Each case includes cap inserts: 150 cap inserts come with cases of 500 and 100 come with cases of 100.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty