Would you like to log in again?
Test Tubes, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CELLSTAR®
115101 115261 184261 169101 120180 187201 151101 160201 187101 187261 163201 136101 191161 160101 163161 115262 186171 188161 187262 163177 191180 112201 163261 164161 188171 120161 103101
89049-102CS 700.65 USD InStock
89049-102 89049-100 10066-770 82050-270 82050-292 82050-138 82050-116 82050-238 82050-150 82050-272 82050-294 82050-274 82050-252 82050-232 82050-256 82050-234 89049-098 82050-268 82050-246 82050-148 82050-104 82050-248 82050-128 82050-228 82050-140 82050-266 82050-244
Test Tubes, Greiner Bio-One
Tubes Culture Tubes
These high-quality test tubes are available in a wide variety of materials, sizes, and volume ranges.
  • Excellent optical clarity
  • Round, flat, or conical bottom designs available
  • Polycarbonate, polypropylene, or polystyrene construction available
  • Exceptional thermal and chemical stability
Test tubes provide excellent optical clarity, and are available in round or conical bottom designs. Unless otherwise noted, tube caps are not included.
Ordering information: Test tube 82050-248 is supplied sterile.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty