Would you like to log in again?
CELLSTAR® Cell Culture Tubes, Disposable, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
CELLSTAR®
191160 163160
82050-290CS 193.61 USD InStock
82050-290 82050-242
CELLSTAR® Cell Culture Tubes, Disposable, Polystyrene, Sterile, Greiner Bio-One
Tubes Culture Tubes
CELLSTAR® Cell culture tubes are made of high-grade polystyrene and are available with screw cap, bayonet cap or two-position vent stopper.
  • Sterile
  • Disposable
  • Free of RNase, DNase, and human DNA
  • Nonpyrogenic
  • Polystyrene construction
The clarity of polystyrene makes tubes ideal for optical measurements.
Screw caps ensure a secure seal. Ventilation snap caps have two positions, allowing either controlled gas exchange or a secure seal. All tubes are supplied sterile, RNase-, DNase-, and human DNA-free, noncytotoxic (EN ISO 10993), and nonpyrogenic.
Tubes offer improved cell adhesion through physical surface treatment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty